missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Pyrimidinecarboximidamide Hydrochloride, TRC

Artikelnummer. 30424966
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30424966 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424966 Supplier Toronto Research Chemicals Supplier No. TRCP997270500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-Pyrimidinecarboximidamide Hydrochloride
CAS 138588-40-6
Molekylær formel C5 H6 N4 . Cl H
Mængde 500 mg
InChI formel InChI=1S/C5H6N4.ClH/c6-4(7)5-8-2-1-3-9-5;/h1-3H,(H3,6,7);1H
SMIL Cl.NC(=N)c1ncccn1
IUPAC navn pyrimidine-2-carboximidamide;hydrochloride
Molekylvægt (g/mol) 158.59
Formel vægt 158.0359
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.