Få mere at vide
(2-Pyridyl)dithiobimane, TRC
Artikelnummer. 30424139
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30424139 10 mg
30422364 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30424139 Leverandør Toronto Research Chemicals Leverandørnr. TRCP99300010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 385398-64-1
Molekylær formel C15 H15 N3 O2 S2
Molekylvægt (g/mol) 333.43
IUPAC navn 1,2,6-trimethyl-7-[(pyridin-2-yldisulfanyl)methyl]pyrazolo[1,2-a]pyrazole-3,5-dione
SMIL CC1=C(C)C(=O)N2N1C(=C(C)C2=O)CSSc3ccccn3
Tekniske data
Kemisk navn eller materiale (2-Pyridyl)dithiobimane
Mængde 10 mg
InChI formel InChI=1S/C15H15N3O2S2/c1-9-11(3)17-12(10(2)15(20)18(17)14(9)19)8-21-22-13-6-4-5-7-16-13/h4-7H,8H2,1-3H3
Formel vægt 333.0606