Få mere at vide
2-Pyridinamine-d4, TRC
Artikelnummer. 30410808
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30410808 50 mg
30422573 10 mg
30424739 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30410808 Leverandør Toronto Research Chemicals Leverandørnr. TRCP99178750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 87802-54-8
Molekylær formel C5H2D4N2
Molekylvægt (g/mol) 98.14
Synonym 2-Aminopyridine-d4, 1,2-Dihydro-2-iminopyridine-d4, 2-Aminopyridine-d4, 2-Pyridinylamine-d4, 2-Pyridylamine-d4, NSC 431-d4, o-Aminopyridine-d4, α-Aminopyridine-d4, α-Pyridinamine-d4, α-Pyridylamine-d4
SMIL [2H]C1=C([2H])C([2H])=C([2H])C(N)=N1
Tekniske data
Kemisk navn eller materiale 2-Pyridinamine-d4
Mængde 50 mg
InChI formel InChI=1S/C5H6N2/c6-5-3-1-2-4-7-5/h1-4H,(H2,6,7)/i1D,2D,3D,4D
Formel vægt 98.0782