missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Pyrazinecarboxylsyre 4-oxid, TRC

Artikelnummer. 30428789
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30428789 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30428789 Leverandør Toronto Research Chemicals Leverandørnr. TRCP84080025MG
 Begrænset vare
Forventet forsendelse: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-Pyrazinecarboxylic Acid 4-Oxide
CAS 874-54-4
Molekylær formel C5H4N2O3
Mængde 25 mg
Synonym Acipimox Impurity 7
InChI formel InChI=1S/C5H4N2O3/c8-5(9)4-3-7(10)2-1-6-4/h1-3H,(H,8,9)
SMIL OC(=O)c1c[n+]([O-])ccn1
IUPAC navn 4-oxidopyrazin-4-ium-2-carboxylsyre
Molekylvægt (g/mol) 140.1
Formel vægt 140.0222
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.