Learn More
2-Propyn-1-ol, Sigma-Aldrich™
Product Code. 15819410
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15819410 1 L 1 L
15809410 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15819410 Supplier Sigma Aldrich Supplier No. 8070501000
 Restricted Item
Forventet forsendelse: 26-11-2026
For synthesis
for synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 107-19-7
Grade Research
Molecular Weight (g/mol) 56.06
Specifications
Melting Point -53°C
Density 0.95 g/cm3 at 20°C
Boiling Point 114°C to 115°C/1013 hPa
MDL Number MFCD00002912
Quantity 1 L
Synonym 2-Propyn-1-ol, Propargyl alcohol, Propinol, Ethynyl carbinol, 2-Propyn-1-ol