Få mere at vide
2-Propylglutaric Acid, TRC
Artikelnummer. 30428411
Skift visning
Mængde:
1 mg
2.5 mg
5 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30428411 1 mg -
30417827 5 mg -
30421680 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30428411 Leverandør Toronto Research Chemicals Leverandørnr. TRCP8353001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 32806-62-5
Molekylær formel C8 H14 O4
Molekylvægt (g/mol) 174.19
IUPAC navn 2-propylpentanedioic acid
SMIL CCCC(CCC(=O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale 2-Propylglutaric Acid
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C8H14O4/c1-2-3-6(8(11)12)4-5-7(9)10/h6H,2-5H2,1H3,(H,9,10)(H,11,12)
Formel vægt 174.0892
Procent renhed >95%