Få mere at vide
(+/-)-2-Propyl-4-pentenoic Acid, TRC
Artikelnummer. 30003593
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30003593 500 mg
17984225 100 mg
17984175 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30003593 Leverandør Toronto Research Chemicals Leverandørnr. TRCP838305500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1575-72-0
Molekylær formel C8 H14 O2
Molekylvægt (g/mol) 142.2
Synonym 4-Pentenoic acid, 2-propyl-, Pentanoic acid, 2-(2-propenyl)-, 2-Propyl-4-pentenoic acid, (±)-2-n-Propyl-4-pentenoic acid, 2-Allylpentanoic acid, 2-n-Propyl-4-pentenoic acid
IUPAC navn 2-propylpent-4-enoic acid
SMIL CCCC(CC=C)C(=O)O
Tekniske data
Kemisk navn eller materiale (+/-)-2-Propyl-4-pentenoic Acid
Mængde 500 mg
InChI formel InChI=1S/C8H14O2/c1-3-5-7(6-4-2)8(9)10/h3,7H,1,4-6H2,2H3,(H,9,10)
Formel vægt 142.0994