Få mere at vide
2-Propyl-2-heptenal (Technical Grade), TRC
Artikelnummer. 30428520
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30428520 1 g
30423919 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30428520 Leverandør Toronto Research Chemicals Leverandørnr. TRCP8358151G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 34880-43-8
Molekylær formel C10H18O
Molekylvægt (g/mol) 154.25
Synonym 2-Propyl-2-hepten-1-al
SMIL CCCCCC(CCC)C=O
Tekniske data
Kemisk navn eller materiale 2-Propyl-2-heptenal (Technical Grade)
Mængde 1 g
InChI formel InChI=1S/C10H18O/c1-3-5-6-8-10(9-11)7-4-2/h8-9H,3-7H2,1-2H3/b10-8+