Få mere at vide
2-(Propyl-2,2-d2)pentanoic-4,4-d2 Acid, CDN
Artikelnummer. 30321196
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321196 0.05 g
30311283 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321196 Leverandør CDN Leverandørnr. CDND28840.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Valproic-4,4,4',4'-D4 Acid
CAS 345909-03-7
Molekylær formel C8 D4 H12 O2
Mængde 0.05 g
Synonym Pentanoic-4,4-d2 acid, 2-(propyl-2,2-d2)- (9 CI), 4,4,4',4'-Tetradeuterovalproic acid, 2-(Propyl-2,2-d2)pentanoic-4,4-d2 acid, Valproic acid-d4, Valproic-4,4,4',4'-D4 acid
InChI formel InChI=1 S/C8H16O2/c1-3-5-7(6-4-2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10)/i3D2,4D2
SMIL [2 H]C([2 H])(C)CC(CC([2 H])([2 H])C)C(=O)O
IUPAC navn 4,4-dideuterio-2-(2,2-dideuteriopropyl)pentanoic acid
Molekylvægt (g/mol) 148.2361
Formel vægt 148.1401 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity