Få mere at vide
2-Piperazinyl-4-amino-6,7-dimethoxyquinazoline-d8, TRC
Artikelnummer. 30423703
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30423703 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30423703 Leverandør Toronto Research Chemicals Leverandørnr. TRCP4805022.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Piperazinyl-4-amino-6,7-dimethoxyquinazoline-d8
CAS 1246820-89-2
Molekylær formel C14H11D8N5O2
Mængde 2.5 mg
Synonym 6,7-Dimethoxy-2-(1-piperazinyl)-4-quinazolinamine-d8, 2-(1-Piperazinyl)-4-amino-6,7-dimethoxyquinazoline-d8, 4-Amino-2-(1-piperazinyl)-6,7-dimethoxyquinazoline-d8, 4-Amino-6,7-dimethoxy-2-(1-piperazinyl)quinazoline-d8
InChI formel InChI=1S/C14H19N5O2/c1-20-11-7-9-10(8-12(11)21-2)17-14(18-13(9)15)19-5-3-16-4-6-19/h7-8,16H,3-6H2,1-2H3,(H2,15,17,18)/i3D2,4D2,5D2,6D2
SMIL [2H]C1([2H])NC([2H])([2H])C([2H])([2H])N(c2nc(N)c3cc(OC)c(OC)cc3n2)C1([2H])[2H]
IUPAC navn 6,7-dimethoxy-2-(2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl)quinazolin-4-amine
Molekylvægt (g/mol) 297.38
Formel vægt 297.2041