Få mere at vide
2-Picoline-d7, TRC
Artikelnummer. 30408981
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30408981 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30408981 Leverandør Toronto Research Chemicals Leverandørnr. TRCP4371971G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Picoline-d7
CAS 93951-93-0
Molekylær formel C6 D7 N
Mængde 1 g
Synonym 2-Methylpyridine-d7, o-Picoline-d7, o-Methylpyridine-d7, ortho-Picoline-d7, ortho-Methylpyridine-d7
InChI formel InChI=1S/C6H7N/c1-6-4-2-3-5-7-6/h2-5H,1H3/i1D3,2D,3D,4D,5D
SMIL [2H]c1nc(c([2H])c([2H])c1[2H])C([2H])([2H])[2H]
IUPAC navn 2,3,4,5-tetradeuterio-6-(trideuteriomethyl)pyridine
Molekylvægt (g/mol) 100.17
Formel vægt 100.1018