Få mere at vide
2-Phenylpropionitrile-d5, TRC
Artikelnummer. 30419350
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30419350 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419350 Leverandør Toronto Research Chemicals Leverandørnr. TRCP33620210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Phenylpropionitrile-d5
CAS 1330238-27-1
Molekylær formel C9H4D5N
Mængde 10 mg
Synonym alpha-Methyl-benzeneacetonitrile-d5, Hydratroponitrile-d5, (+/-)-alpha-Methylbenzeneacetonitrile-d5, NSC 11264-d5, NSC 37485-d5, dl-2-Phenylpropionitrile-d5, dl-alpha-Phenylethyl Isocyanide-d5, alpha-Cyanoethylbenzene-d5, alpha-Methylbenzeneacetonitrile-d5, alpha-Methylbenzyl Cyanide-d5, alpha-Methylphenylacetonitrile-d5, alpha-Phenylethyl Cyanide-d5
InChI formel InChI=1S/C9H9N/c1-8(7-10)9-5-3-2-4-6-9/h2-6,8H,1H3/i2D,3D,4D,5D,6D
SMIL [2H]c1c([2H])c([2H])c(C(C)C#N)c([2H])c1[2H]
IUPAC navn 2-(2,3,4,5,6-pentadeuteriophenyl)propanenitrile
Molekylvægt (g/mol) 136.21
Formel vægt 136.1049