Få mere at vide
2-Phenylpropane-2-d1, CDN
Artikelnummer. 30320236
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320236 0.25 g -
30324380 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320236 Leverandør CDN Leverandørnr. CDND06790.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Isopropylbenzene-alpha-D
CAS 4019-54-9
Molekylær formel C9 D H11
Mængde 0.25 g
Synonym Benzene, (1-methylethyl-1-d)- (9 CI, ACI), (1-Methylethyl-1-d)benzene (ACI), Cumene-α-d (6 CI, 7 CI, 8 CI), 2-Deutero-2-phenylpropane, Dimethylphenylmethane-d1, Isopropylbenzene-d, Isopropylbenzene-α-D, 2-Phenylpropane-2-d1
InChI formel InChI=1 S/C9H12/c1-8(2)9-6-4-3-5-7-9/h3-8 H,1-2H3/i8D
SMIL [2 H]C(C)(C)c1ccccc1
IUPAC navn 2-deuteriopropan-2-ylbenzene
Molekylvægt (g/mol) 121.1977
Formel vægt 121.1002 g/mol
Procent renhed 97 atom % D, min 98% Chemical Purity