Få mere at vide
2-Phenylpropane-1,1,1,3,3,3-d6, CDN
Artikelnummer. 30321598
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321598 0.1 g
30313803 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321598 Leverandør CDN Leverandørnr. CDND06800.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Isopropylbenzene-D6 (dimethyl-D6)
CAS 20201-29-0
Molekylær formel C9 D6 H6
Mængde 0.1 g
Synonym Benzene, [1-(methyl-d3)ethyl-2,2,2-d3]- (9 CI, ACI), [1-(Methyl-d3)ethyl-2,2,2-d3]benzene (ACI), Cumene-β,β,β,β',β',β'-d6 (8 CI), Isopropylbenzene-d6, Isopropylbenzene-D6 (dimethyl-D6), 2-Phenylpropane-1,1,1,3,3,3-d6
InChI formel InChI=1 S/C9H12/c1-8(2)9-6-4-3-5-7-9/h3-8 H,1-2H3/i1D3,2D3
SMIL [2 H]C([2 H])([2 H])C(c1ccccc1)C([2 H])([2 H])[2 H]
IUPAC navn 1,1,1,3,3,3-hexadeuteriopropan-2-ylbenzene
Molekylvægt (g/mol) 126.2286
Formel vægt 126.1316 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity