missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Phenylbutyric Acid-d5 Methyl Ester, TRC

Artikelnummer. 30426057
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30426057 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30426057 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3196075MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2-Phenylbutyric Acid-d5 Methyl Ester
CAS 1794940-35-4
Molekylær formel C11H9D5O2
Mængde 5 mg
Synonym 2-(Ethyl-d5)-2-phenylacetic Acid Methyl Ester, alpha-(Ethyl-d5)benzeneacetic Acid Methyl Ester, (RS)-2-Phenylbutanoic Acid-d5 Methyl Ester, (+/-)-2-Phenylbutanoic Acid-d5 Methyl Ester, alpha-(Ethyl-d5)-alpha-toluic Acid Methyl Ester, alpha-(Ethyl-d5)benzeneacetic Acid Methyl Ester, alpha-(Ethyl-d5)phenylacetic Acid Methyl Ester
InChI formel InChI=1S/C11H14O2/c1-3-10(11(12)13-2)9-7-5-4-6-8-9/h4-8,10H,3H2,1-2H3/i1D3,3D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C(C(=O)OC)c1ccccc1
IUPAC navn methyl 3,3,4,4,4-pentadeuterio-2-phenylbutanoate
Molekylvægt (g/mol) 183.26
Formel vægt 183.1308
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.