Få mere at vide
2-Phenylbicyclo[2.2.1]heptane-2-carboxylic Acid, TRC
Artikelnummer. 30425131
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30425131 10 mg -
30428603 2.5 mg 2.5mg
30425962 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30425131 Leverandør Toronto Research Chemicals Leverandørnr. TRCP29785510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 93963-31-6
Molekylær formel C14H16O2
Molekylvægt (g/mol) 216.28
Synonym Bornaprine Impurity 1
IUPAC navn 3-phenylbicyclo[2.2.1]heptane-3-carboxylic acid
SMIL OC(=O)C1(CC2CCC1C2)c3ccccc3
Tekniske data
Kemisk navn eller materiale 2-Phenylbicyclo[2.2.1]heptane-2-carboxylic Acid
Mængde 10 mg
InChI formel InChI=1S/C14H16O2/c15-13(16)14(11-4-2-1-3-5-11)9-10-6-7-12(14)8-10/h1-5,10,12H,6-9H2,(H,15,16)
Formel vægt 216.115