Learn More
2-Phenylbenzimidazole-5-sulfonic Acid, 98%, BLD Pharm™
Product Code. 30948018
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30948018 500 g 500g
30930545 25 g 25g
30934497 100 g 100g
30938992 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30948018 Supplier BLD Pharm Supplier No. BD14523500g
 Restricted Item
Forventet forsendelse: 27-11-2026
Specifications
CAS 27503-81-7
Molecular Formula C13H10N2O3S
MDL Number MFCD00053007
Quantity 500 g
SMILES O=S(C1=CC=C2C(NC(C3=CC=CC=C3)=N2)=C1)(O)=O
Molecular Weight (g/mol) 274.30
Percent Purity 98%
Chemical Name or Material 2-Phenylbenzimidazole-5-sulfonic Acid