Få mere at vide
2-Phenyl-d5-ethan-1,1,2,2-d4-ol, CDN
Artikelnummer. 30320264
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320264 0.1 g
30319863 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320264 Leverandør CDN Leverandørnr. CDND54800.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Phenylethanol-D9
CAS 42950-74-3
Molekylær formel C8 D9 H O
Mængde 0.1 g
Synonym Benzene-d5-ethan-d4-ol (9 CI), Benzene-d5-ethan-α,α,β,β-d4-ol, 2-Phenyl-d5-ethan-1,1,2,2-d4-ol, 2-(2,3,4,5,6-Pentadeuteriophenyl)-1,1,2,2-tetradeuterioethanol, 2-(Phenyl-d5)ethanol-1,1,2,2-d4, Phenethyl Alcohol-d9, Phenethyl-d9 alcohol, 2-Phenylethanol-D9
InChI formel InChI=1 S/C8H10O/c9-7-6-8-4-2-1-3-5-8/h1-5,9 H,6-7H2/i1D,2 D,3 D,4 D,5 D,6D2,7D2
SMIL [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])C([2 H])([2 H])O
IUPAC navn 1,1,2,2-tetradeuterio-2-(2,3,4,5,6-pentadeuteriophenyl)ethanol
Molekylvægt (g/mol) 131.2199
Formel vægt 131.1297 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity