Få mere at vide
2-Phenyl-d5-4,4-dimethyl-4,5-dihydrooxazole, TRC
Artikelnummer. 30414557
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30414557 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30414557 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3197375MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Phenyl-d5-4,4-dimethyl-4,5-dihydrooxazole
CAS 639516-58-8
Molekylær formel C11 2H5 H8 N O
Mængde 5 mg
InChI formel InChI=1S/C11H13NO/c1-11(2)8-13-10(12-11)9-6-4-3-5-7-9/h3-7H,8H2,1-2H3/i3D,4D,5D,6D,7D
SMIL [2H]c1c([2H])c([2H])c(C2=NC(C)(C)CO2)c([2H])c1[2H]
IUPAC navn 4,4-dimethyl-2-(2,3,4,5,6-pentadeuteriophenyl)-5H-1,3-oxazole
Molekylvægt (g/mol) 180.26
Formel vægt 180.1311