Få mere at vide
(2-Phenyl-1H-imidazol-4-yl)methanol, TRC
Artikelnummer. 30421570
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30421570 500 mg
30421135 250 mg
30410251 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30421570 Leverandør Toronto Research Chemicals Leverandørnr. TRCP229425500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 43002-54-6
Molekylær formel C10H10N2O
Molekylvægt (g/mol) 174.2
IUPAC navn (2-phenyl-1H-imidazol-5-yl)methanol
SMIL OCc1cnc([nH]1)c2ccccc2
Tekniske data
Kemisk navn eller materiale (2-Phenyl-1H-imidazol-4-yl)methanol
Mængde 500 mg
InChI formel InChI=1S/C10H10N2O/c13-7-9-6-11-10(12-9)8-4-2-1-3-5-8/h1-6,13H,7H2,(H,11,12)
Formel vægt 174.0793