Få mere at vide
2-Phenoxyethyl-1,1,2,2-d4 Alcohol, TRC
Artikelnummer. 30416109
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30416109 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30416109 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2979015MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(Phenoxy)ethanol-1,1,2,2-D4
CAS 1219804-65-5
Molekylær formel C8H6D4O2
Mængde 5 mg
Synonym 2-Phenoxyethyl-1,1,2,2-d4 Alcohol
InChI formel InChI=1S/C8H10O2/c9-6-7-10-8-4-2-1-3-5-8/h1-5,9H,6-7H2/i6D2,7D2
SMIL OC([2H])(C([2H])(OC1=CC=CC=C1)[2H])[2H]
IUPAC navn 2-phenoxyethan-1,1,2,2-d4-1-ol
Molekylvægt (g/mol) 142.19
Formel vægt 142.09