Få mere at vide
2-Phenoxyethanol-d2, TRC
Artikelnummer. 30409327
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30409327 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30409327 Leverandør Toronto Research Chemicals Leverandørnr. TRCP29790310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Phenoxyethanol-d2
CAS 21273-38-1
Renhedsgrad noter GC
Molekylær formel C8 D2 H8 O2
Mængde 10 mg
Synonym Ethan-1,1-d2-ol','2-phenoxy- (8CI,9CI), Phenoxyethanol-d2, (2-Hydroxyethoxy-2,2-d2)benzene, β-Hydroxyethyl-d2 phenyl ether, β-Phenoxyethanol-d2, 1-Hydroxy-2-phenoxyethane-1,1-d2, 2-Hydroxyethyl-d2 phenyl ether, 2-Phenoxyethanol-d2, 2-Phenoxyethyl-1,1-d2 alcohol, 2-(Phenoxy)ethanol-1,1-D2
InChI formel InChI=1S/C8H10O2/c9-6-7-10-8-4-2-1-3-5-8/h1-5,9H,6-7H2/i6D2
SMIL [2H]C([2H])(O)COc1ccccc1
IUPAC navn 1,1-dideuterio-2-phenoxyethanol
Molekylvægt (g/mol) 140.176
Formel vægt 140.081
Procent renhed >95