Få mere at vide
2-Phenanthrol-d9 (Major), TRC
Artikelnummer. 30422907
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30422907 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422907 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2950071MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Phenanthrol-d9
CAS 922510-19-8
Molekylær formel C14HD9O
Mængde 1 mg
Synonym 2-Phenanthrenol-d9, 2-Hydroxyphenanthrene-d9, NSC 2576-d9
InChI formel InChI=1S/C14H10O/c15-12-7-8-14-11(9-12)6-5-10-3-1-2-4-13(10)14/h1-9,15H/i1D,2D,3D,4D,5D,6D,7D,8D,9D
SMIL OC(C([2H])=C1[2H])=C([2H])C2=C1C3=C([2H])C([2H])=C([2H])C([2H])=C3C([2H])=C2[2H]
Molekylvægt (g/mol) 203.28