Få mere at vide
2-Perfluoropropoxy-2,3,3,3-tetrafluoropropanol, TRC
Artikelnummer. 30412089
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30412089 500 mg
30421334 100 mg
30419826 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30412089 Leverandør Toronto Research Chemicals Leverandørnr. TRCP999433500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 26537-88-2
Molekylær formel C6H3F11O2
Molekylvægt (g/mol) 316.07
IUPAC navn 2,3,3,3-tetrafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propan-1-ol
SMIL OCC(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F
Tekniske data
Kemisk navn eller materiale 2-Perfluoropropoxy-2,3,3,3-tetrafluoropropanol
Mængde 500 mg
InChI formel InChI=1S/C6H3F11O2/c7-2(1-18,4(10,11)12)19-6(16,17)3(8,9)5(13,14)15/h18H,1H2
Formel vægt 315.9957