Få mere at vide
2-Pentanone-1,1,1,3,3-d5, CDN
Artikelnummer. 30315949
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30315949 0.5 g 0.5g
30314201 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315949 Leverandør CDN Leverandørnr. CDND02190.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Pentanone D5 (1,1,1,3,3-D5)
CAS 24313-49-3
Molekylær formel C5 D5 H5 O
Mængde 0.5 g
Synonym 2-Pentanone-1,1,1,3,3-d5, 1,1,1,3,3-Pentadeuterio-2-pentanone, 2-Pentanone-1,1,1,3,3-D5, Pentan-2-one-d5
InChI formel InChI=1 S/C5H10O/c1-3-4-5(2)6/h3-4H2,1-2H3/i2D3,4D2
SMIL [2 H]C([2 H])([2 H])C(=O)C([2 H])([2 H])CC
IUPAC navn 1,1,1,3,3-pentadeuteriopentan-2-one
Molekylvægt (g/mol) 91.1631
Formel vægt 91.1045 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity