Få mere at vide
2-Oxohexanoic Acid Sodium Salt, TRC
Artikelnummer. 30422764
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30422764 25 mg
30411136 100 mg
30411810 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30422764 Leverandør Toronto Research Chemicals Leverandørnr. TRCO91308525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13022-85-0
Molekylær formel C6H10NaO3
Molekylvægt (g/mol) 152.12
Synonym Sodium 2-oxohexanoate, 2-Ketocaproic Acid Sodium Salt
IUPAC navn sodium;2-oxohexanoate
SMIL [Na+].CCCCC(=O)C(=O)[O-]
Tekniske data
Kemisk navn eller materiale 2-Oxohexanoic Acid Sodium Salt
Mængde 25 mg
InChI formel InChI=1S/C6H10O3.Na/c1-2-3-4-5(7)6(8)9;/h2-4H2,1H3,(H,8,9);/q;+1/p-1
Formel vægt 152.0449