Få mere at vide
2’-Oxo Ifosfamide-d4, TRC
Artikelnummer. 30424207
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30424207 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30424207 Leverandør Toronto Research Chemicals Leverandørnr. TRCO85695210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2'-Oxo Ifosfamide-d4
CAS 1329497-14-4
Molekylær formel C7H9D4Cl2N2O3P
Mængde 10 mg
Synonym 2-Chloro-1-[2-[(2-chloroethyl)amino]dihydro-2-oxido-2H-1,3,2-oxazaphosphorin-3(4H)-yl]ethanone-d4, 3-(Chloroacetyl)-N-(2-chloroethyl)tetrahydro-2H-1,3,2-oxazaphosphorin-2-amine-d4 2-Oxide
InChI formel InChI=1S/C7H13Cl2N2O3P/c8-2-3-10-15(13)11(7(12)6-9)4-1-5-14-15/h1-6H2,(H,10,13)/i1D2,4D2
SMIL [2H]C1(COP(=O)(N(C1([2H])[2H])C(=O)CCl)NCCCl)[2H]
Molekylvægt (g/mol) 279.09
Formel vægt 278.0292