missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Oxo Clopidogrel Hydrochloride (Mixture of Diastereomers), TRC

Artikelnummer. 30415853
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30415853 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30415853 Leverandør Toronto Research Chemicals Leverandørnr. TRCO8699905MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2-Oxo Clopidogrel Hydrochloride (Mixture of Diastereomers)
CAS 1219432-42-4
Molekylær formel C16H17Cl2NO3S
Mængde 5 mg
Synonym α-(2-Chlorophenyl)-2,6,7,7a-tetrahydro-2-oxo-thieno[3,2-c]pyridine-5(4H)-acetic Acid Methyl Ester Hydrochloride
InChI formel InChI=1S/C16H16ClNO3S.ClH/c1-21-16(20)15(11-4-2-3-5-12(11)17)18-7-6-13-10(9-18)8-14(19)22-13;/h2-5,8,13,15H,6-7,9H2,1H3;1H
SMIL ClC1=C(C(C(OC)=O)N2CCC3C(C2)=CC(S3)=O)C=CC=C1.Cl
Molekylvægt (g/mol) 374.28
Formel vægt 373.0306
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.