Få mere at vide
2-Oxo-3-methylbutanoic acid >90%, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-Oxo-3-methylbutanoic acid |
| CAS | 759-05-7 |
| Molekylær formel | C5H8O3 |
| Mængde | 100 mg |
| Synonym | Butanoic acid','3-methyl-2-oxo-, Butyric acid','3-methyl-2-oxo- (8CI), 3-Methyl-2-oxobutanoic acid, 2-Ketoisovaleric acid, 2-Oxo-3-methylbutanoic acid, 2-Oxo-3-methylbutyric acid, 2-Oxoisovaleric acid, 2-keto-3-Methylbutyric acid, 2-keto-Isovaleric acid, 3-Methyl-2-oxobutyrate, 3-Methyl-2-oxobutyric acid, Dimethylpyruvic acid, Isopropylglyoxylic acid, Ketovaline, α-Ketoisovaleric acid, α-Oxo-β-methylbutyric acid, α-Oxoisovaleric acid, α-keto-Isovaleric acid, α-keto-β-Methylbutyric acid |
| InChI formel | InChI=1S/C5H8O3/c1-3(2)4(6)5(7)8/h3H,1-2H3,(H,7,8) |
| SMIL | CC(C)C(=O)C(=O)O |
| IUPAC navn | 3-methyl-2-oxobutanoic acid |
| Molekylvægt (g/mol) | 116.12 |
| Formel vægt | 116.0473 |