missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
2-Oxo-3-hydroxy-LSD, Lipomed
Description
2-Oxo-3-hydroxy-LSD
Specifications
Specifications
| Kemisk navn eller materiale | 2-Oxo-3-hydroxy-LSD |
| CAS | 111295-09-1 |
| Analyt- eller komponentnavne | 2-Oxo-3-hydroxy-LSD |
| Forsendelsestilstand | Stuetemperatur |
| Molekylær formel | C20 H25 N3 O3 |
| Synonym | 2-Oxo-3-hydroxy-LSD, 2,3-Dihydro-3-hydroxy-2-oxo Lysergide |
| InChI formel | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMIL | CCN(CC)C(=O)[C@H]1 CN(C)[C@@H]2 CC3(O)C(=O)Nc4cccc(C2=C1)c34 |
| IUPAC navn | (6 aR,9 R)-N,N-diethyl-5 A-hydroxy-7-methyl-5-oxo-6,6 A,8,9-tetrahydro-4 H-indolo[4,3-fg]kinolin-9-carboxamid |
| Industry Type | Farmaceutisk |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?