Få mere at vide
2-Oxo-2-[[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]amino]ethyl Pentanoate, TRC
Artikelnummer. 30428450
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30428450 10 mg
30425417 5 mg
30417038 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30428450 Leverandør Toronto Research Chemicals Leverandørnr. TRCO43212510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 914465-61-5
Molekylær formel C21 H23 N5 O3
Molekylvægt (g/mol) 393.44
Synonym Pentanoic acid, 2-oxo-2-[[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]amino]ethyl ester, Pentanoic acid, 2-oxo-2-[[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]amino]ethyl ester (9CI)
IUPAC navn [2-oxo-2-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methylamino]ethyl] pentanoate
SMIL CCCCC(=O)OCC(=O)NCc1ccc(cc1)c2ccccc2c3nnn[nH]3
Tekniske data
Kemisk navn eller materiale 2-Oxo-2-(((2'-(2H-tetrazol-5-yl)(1,1'-biphenyl)-4-yl)methyl)amino)ethyl Pentanoate
Mængde 10 mg
InChI formel InChI=1S/C21H23N5O3/c1-2-3-8-20(28)29-14-19(27)22-13-15-9-11-16(12-10-15)17-6-4-5-7-18(17)21-23-25-26-24-21/h4-7,9-12H,2-3,8,13-14H2,1H3,(H,22,27)(H,23,24,25,26)
Formel vægt 393.1801