Få mere at vide
2-Oxo-1,5-pentanedioic Acid-d6, CDN
Artikelnummer. 30324909
Skift visning
Mængde:
0.01 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30324909 0.01 g
30310234 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324909 Leverandør CDN Leverandørnr. CDND81050.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Oxopentanedioic Acid-D6
CAS 1173021-86-7
Molekylær formel C5 D6 O5
Mængde 0.01 g
Synonym Pentanedioic-2,2,3,3-d4 acid-1,5-d2, 4-oxo- (ACI), 4-Oxopentanedioic-2,2,3,3-d4 acid-1,5-d2 (ACI), 2-Ketopentanedioic acid-d6, 2-Oxo-1,5-pentanedioic Acid-d6, 2-Ketoglutaric Acid-d6, 2-Oxopentanedioic-3,3,4,4-d4 acid-d2, α-Ketoglutaric Acid-d6, 2-Oxoglutaric Acid-d6, 2-Oxopentanedioic Acid-D6
InChI formel InChI=1 S/C5H6O5/c6-3(5(9)10)1-2-4(7)8/h1-2H2,(H,7,8)(H,9,10)/i1D2,2D2/hD2
SMIL [2 H]OC(=O)C(=O)C([2 H])([2 H])C([2 H])([2 H])C(=O)O[2 H]
IUPAC navn dideuterio 2,2,3,3-tetradeuterio-4-oxopentanedioate
Molekylvægt (g/mol) 152.1351
Formel vægt 152.0592 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity