missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Oleoyl Glycerol-d5, TRC

Artikelnummer. 30426498
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30426498 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30426498 Leverandør Toronto Research Chemicals Leverandørnr. TRCO5280221MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2-Oleoyl Glycerol-d5
CAS 946524-37-4
Molekylær formel C21H35D5O4
Mængde 1 mg
InChI formel InChI=1S/C21H40O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-20(18-22)19-23/h9-10,20,22-23H,2-8,11-19H2,1H3/b10-9-/i18D2,19D2,20D
SMIL [2H]C([2H])(O)C([2H])(OC(=O)CCCCCCC\C=C/CCCCCCCC)C([2H])([2H])O
IUPAC navn (1,1,2,3,3-pentadeuterio-1,3-dihydroxypropan-2-yl) (Z)-octadec-9-enoate
Molekylvægt (g/mol) 361.57
Formel vægt 361.324
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.