Få mere at vide
2'-O-Methyl Adenosine-d3, TRC
Artikelnummer. 30483484
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483484 2.5 mg
30473826 10 mg
30471450 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30483484 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2761522.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1195030-24-0
Molekylær formel C11 D3 H12 N5 O4
Molekylvægt (g/mol) 284.286
Synonym 2'-O-(Methyl-D3)-Adenosine, 2'-O-Methyl-D-Adenosine-D3, 5-(6-Aminopurin-9-Yl)-2-(Hydroxymethyl)-4-Methoxyoxolan-3-Ol-D3
IUPAC navn (2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)-4-(trideuteriomethoxy)oxolan-3-ol
SMIL [2H]C([2H])([2H])O[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1n2cnc3c(N)ncnc23
Tekniske data
Kemisk navn eller materiale 2'-O-Methyl Adenosine-D3
Mængde 2.5 mg
InChI formel InChI=1S/C11H15N5O4/c1-19-8-7(18)5(2-17)20-11(8)16-4-15-6-9(12)13-3-14-10(6)16/h3-5,7-8,11,17-18H,2H2,1H3,(H2,12,13,14)/t5-,7-,8-,11-/m1/s1/i1D3
Formel vægt 284.131