Få mere at vide
2'-O-beta-D-Ribofuranosylguanosine, TRC
Artikelnummer. 30009264
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009264 5 mg
30005168 25 mg
30003474 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009264 Leverandør Toronto Research Chemicals Leverandørnr. TRCR2233405MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 148711-49-3
Molekylær formel C15 H21 N5 O9
Molekylvægt (g/mol) 415.355
IUPAC navn 2-amino-9-[(2R,3R,4R,5R)-3-[(2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMIL NC1=NC(=O)c2ncn([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O[C@@H]4O[C@H](CO)[C@@H](O)[C@H]4O)c2N1
Tekniske data
Kemisk navn eller materiale 2'-O-beta-D-Ribofuranosylguanosine
Mængde 5 mg
InChI formel InChI=1S/C15H21N5O9/c16-15-18-11-6(12(26)19-15)17-3-20(11)13-10(8(24)5(2-22)27-13)29-14-9(25)7(23)4(1-21)28-14/h3-5,7-10,13-14,21-25H,1-2H2,(H3,16,18,19,26)/t4-,5-,7-,8-,9-,10-,13-,14+/m1/s1
Formel vægt 415.134