Få mere at vide
2'-O-(2-Methoxyethyl)-5-methyl-uridine, TRC
Artikelnummer. 30469838
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30469838 100 mg
30477346 250 mg
30477593 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30469838 Leverandør Toronto Research Chemicals Leverandørnr. TRCM304495100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 163759-49-7
Molekylær formel C13H20N2O7
Molekylvægt (g/mol) 316.31
IUPAC navn 1-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-(2-methoxyethoxy)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
SMIL COCCO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N2C=C(C)C(=O)NC2=O
Tekniske data
Kemisk navn eller materiale 2'-O-(2-Methoxyethyl)-5-methyl-uridine
Mængde 100 mg
InChI formel InChI=1S/C13H20N2O7/c1-7-5-15(13(19)14-11(7)18)12-10(21-4-3-20-2)9(17)8(6-16)22-12/h5,8-10,12,16-17H,3-4,6H2,1-2H3,(H,14,18,19)/t8-,9-,10-,12-/m1/s1
Formel vægt 316.1271