Beskrivelse
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-NP-SCA |
| CAS | 16004-43-6 |
| CAS Max % | ≤ 100% |
| Farve | Colorless |
| Smeltepunkt | 240°C to 245°C |
| Synonym | 4-(2-Nitrobenzylidene)semicarbazide |
| Molekylvægt (g/mol) | 208.18 g/mol |
| Mængde | 10 mg |