Få mere at vide
2-NP-AOZ, TRC
Artikelnummer. 30427234
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30427234 100 mg
30409889 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30427234 Leverandør Toronto Research Chemicals Leverandørnr. TRCN895000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-NP-AOZ
CAS 19687-73-1
Renhedsgrad noter HPLC
Molekylær formel C10 H9 N3 O4
Mængde 100 mg
Synonym 2-Oxazolidinone','3-[(o-nitrobenzylidene)amino]- (8CI), 3-[[(2-Nitrophenyl)methylene]amino]-2-oxazolidinone, NPAOZ
InChI formel InChI=1S/C10H9N3O4/c14-10-12(5-6-17-10)11-7-8-3-1-2-4-9(8)13(15)16/h1-4,7H,5-6H2/b11-7+
SMIL [O-][N+](=O)c1ccccc1\C=N\N2CCOC2=O
IUPAC navn 3-[(E)-(2-nitrophenyl)methylideneamino]-1,3-oxazolidin-2-one
Molekylvægt (g/mol) 235.2
Formel vægt 235.0593
Procent renhed >95