Beskrivelse
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-NP-AOZ |
| CAS | 19687-73-1 |
| CAS Max % | ≤ 100% |
| Smeltepunkt | 186°C to 187°C |
| Synonym | 3-(2-Nitrobenzylidenamino)-2-oxazolidinone |
| Molekylvægt (g/mol) | 235.20 g/mol |
| Mængde | 10 mg |
| Grad | Nitrofuran Metabolites |