Få mere at vide
2-NP-AMOZ-d5, TRC
Artikelnummer. 30417379
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30417379 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30417379 Leverandør Toronto Research Chemicals Leverandørnr. TRCN8949521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-NP-AMOZ D5
CAS 1173097-59-0
Molekylær formel C15 2H5 H13 N4 O5
Mængde 1 mg
Synonym 2-NP-AMOZ-d5, 4,4,5-Trideuterio-5-(dideuterio-morpholin-4-ylmethyl)-3-[(2-nitrophenyl)methylideneamino]-1,3-oxazolidin-2-one, 2-NP-AMOZ D5
InChI formel InChI=1S/C15H18N4O5/c20-15-18(11-13(24-15)10-17-5-7-23-8-6-17)16-9-12-3-1-2-4-14(12)19(21)22/h1-4,9,13H,5-8,10-11H2/b16-9+/i10D2,11D2,13D
SMIL [2H]C([2H])(N1CCOCC1)C2([2H])OC(=O)N(\N=C\c3ccccc3[N+](=O)[O-])C2([2H])[2H]
IUPAC navn 4,4,5-trideuterio-5-[dideuterio(morpholin-4-yl)methyl]-3-[(E)-(2-nitrophenyl)methylideneamino]-1,3-oxazolidin-2-one
Molekylvægt (g/mol) 339.36
Formel vægt 339.1591