Beskrivelse
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-NP-AMOZ-d5 |
| Emballage | Glass Bottle |
| CAS | 1173097-59-0 |
| Farve | White |
| Smeltepunkt | 144°C to 147°C |
| Synonym | 5-(Morpholinomethyl)-3-(2-nitrobenzylidenamino)-2-oxazolidinone-d5 |
| Molekylvægt (g/mol) | 339.36 |
| Mængde | 10 mg |
| Grad | Analytical Standard |