missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Nitropyridine, 97%

Brand:  Alfa Aesar™ H33657.03

Additional Details : CAS Number : 15009-91-3 Weight : 0.00100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15446507

  • 271.00 DKK / 1g

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

15009-91-3
124.099
HLTDBMHJSBSAOM-UHFFFAOYSA-N
26998
C1=CC=NC(=C1)[N+](=O)[O-]
C5H4N2O2
MFCD00160414
pyridine, 2-nitro, nitropyridine, pyridine, nitro, 2-nitro-pyridine, pyridine, 2-nitro-6ci, 7ci, 8ci, 9ci, nitro-pyridine, pyridine, 2-nitro-8ci 9ci, 2-nitropyridine
2-nitropyridine

Specifications

2-Nitropyridine
1g
C5H4N2O2
UN2811
HLTDBMHJSBSAOM-UHFFFAOYSA-N
2-nitropyridine
26998
97%
69°C to 73°C
15009-91-3
MFCD00160414
pyridine, 2-nitro, nitropyridine, pyridine, nitro, 2-nitro-pyridine, pyridine, 2-nitro-6ci, 7ci, 8ci, 9ci, nitro-pyridine, pyridine, 2-nitro-8ci 9ci, 2-nitropyridine
C1=CC=NC(=C1)[N+](=O)[O-]
124.099
124.1
Videos
SDS Documents