Få mere at vide
2-Nitropropane-1,1,1,3,3,3-d6, CDN
Artikelnummer. 30319452
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319452 0.1 g
30314344 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319452 Leverandør CDN Leverandørnr. CDND53930.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Nitropropane-1,1,1,3,3,3-D6
CAS 52809-86-6
Molekylær formel C3 D6 H N O2
Mængde 0.1 g
Synonym Propane-1,1,1,3,3,3-d6, 2-nitro- (9 CI), 1,1,1,3,3,3-Hexadeuterol-2-nitropropane, 2-Nitropropane-1,1,1,3,3,3-d6, 2-Nitropropane-1,1,1,3,3,3-D6, 2-Nitropropane-d6
InChI formel InChI=1 S/C3H7NO2/c1-3(2)4(5)6/h3H,1-2H3/i1D3,2D3
SMIL [2 H]C([2 H])([2 H])C([N+](=O)[O-])C([2 H])([2 H])[2 H]
IUPAC navn 1,1,1,3,3,3-hexadeuterio-2-nitropropane
Molekylvægt (g/mol) 95.1302
Formel vægt 95.0853 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity