Få mere at vide
(2-Nitrophenyl)phenylamine, TRC
Artikelnummer. 30469151
Skift visning
Mængde:
5 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30469151 100 g
30477717 5 g
30470192 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30469151 Leverandør Toronto Research Chemicals Leverandørnr. TRCN504458100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 119-75-5
Molekylær formel C12 H10 N2 O2
Molekylvægt (g/mol) 214.22
Synonym Benzenamine, 2-nitro-N-phenyl- (9CI, ACI), 2-Nitro-N-phenylbenzenamine (ACI), Diphenylamine, 2-nitro- (8CI), 2-Nitro-N-phenylaniline, 2-Nitrodiphenylamine, C.I. 10335, N-(2-Nitrophenyl)aniline, N-Phenyl-2-nitroaniline, N-Phenyl-2-nitrobenzenamine, N-Phenyl-o-nitroaniline, NSC 105613, NSC 4754, NSC 629274, o-(Phenylamino)nitrobenzene, o-Nitro-N-phenylaniline, o-Nitrodiphenylamine, Sudan Yellow 1339
IUPAC navn 2-nitro-N-phenylaniline
SMIL [O-][N+](=O)c1ccccc1Nc2ccccc2
Tekniske data
Kemisk navn eller materiale 2-Nitrodiphenylamine
Mængde 100 g
InChI formel InChI=1S/C12H10N2O2/c15-14(16)12-9-5-4-8-11(12)13-10-6-2-1-3-7-10/h1-9,13H
Formel vægt 214.0742