Få mere at vide
2-Nitrophenol-3,4,5,6-d4, TRC
Artikelnummer. 30473778
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30473778 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473778 Leverandør Toronto Research Chemicals Leverandørnr. TRCN4969065MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Nitrophenol-d4
CAS 93951-78-1
Molekylær formel C6 D4 H N O3
Mængde 5 mg
Synonym Phen-2,3,4,5-d4-ol','6-nitro- (9CI), 2-Nitrophenol-D4, 2-Hydroxy-3,4,5,6-tetradeuterionitrobenzene, o-Hydroxynitrobenzene-d4, o-Nitrophenol-d4, 2-Nitrophenol-3,4,5,6-d4
InChI formel InChI=1S/C6H5NO3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c([2H])c(c(O)c1[2H])[N+](=O)[O-]
IUPAC navn 2,3,4,5-tetradeuterio-6-nitrophenol
Molekylvægt (g/mol) 143.1334
Formel vægt 143.0521