missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrofluorene, 98%, Thermo Scientific Chemicals

Catalog Number p-7052777
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
607-57-8
C13H9NO2
211.22
MFCD00001117
XFOHWECQTFIEIX-UHFFFAOYSA-N
2-nitrofluorene, nitrofluorene, 9h-fluorene, 2-nitro, fluorene, 2-nitro, unii-191ll4u4gz, ccris 1189, dsstox_cid_971, dsstox_rid_75898, dsstox_gsid_20971
11831
CHEBI:1224
2-nitro-9H-fluorene
[O-][N+](=O)C1=CC=C2C(CC3=CC=CC=C23)=C1
This item is not returnable. View return policy
607-57-8
C13H9NO2
211.22
MFCD00001117
XFOHWECQTFIEIX-UHFFFAOYSA-N
2-nitrofluorene, nitrofluorene, 9h-fluorene, 2-nitro, fluorene, 2-nitro, unii-191ll4u4gz, ccris 1189, dsstox_cid_971, dsstox_rid_75898, dsstox_gsid_20971
11831
CHEBI:1224
2-nitro-9H-fluorene
[O-][N+](=O)C1=CC=C2C(CC3=CC=CC=C23)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 156°C to 158°C
Color White to Brown
Quantity 5 g
UN Number UN3077
Formula Weight 211.22
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Nitrofluorene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'