Få mere at vide
2-Nitrobiphenyl-2',3',4',5',6'-d5, TRC
Artikelnummer. 30475831
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30475831 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30475831 Leverandør Toronto Research Chemicals Leverandørnr. TRCN49578850MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Nitrobiphenyl-2',3',4',5',6'-d5
CAS 64420-97-9
Molekylær formel C12H4D5NO2
Mængde 50 mg
Synonym 1,1'-Biphenyl-2,3,4,5,6-d5','2'-nitro- (9CI), 2-Nitrobiphenyl-2',3',4',5',6'-d5, 2-Nitrobiphenyl-2',3',4',5',6'-D5, 2-Nitrobiphenyl-d5
InChI formel InChI=1S/C12H9NO2/c14-13(15)12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9H/i1D,2D,3D,6D,7D
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])c2ccccc2[N+](=O)[O-]
Molekylvægt (g/mol) 204.24
Formel vægt 204.0947