missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nitrobenzyl alcohol, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
The o-nitrobenzyl group (o-NB) is utilized frequently in polymer and materials science. It covers the use of based cross-linkers for photodegradable hydrogels, o-NB side chain functionalization in (block) copolymers, o-NB side chain functionalization for thin film patterning, o-NB for self-assembled monolayers, photo cleavable block copolymers, and photo cleavable bioconjugates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The o-nitrobenzyl group (o-NB) is utilized frequently in polymer and materials science. It covers the use of based cross-linkers for photodegradable hydrogels, o-NB side chain functionalization in (block) copolymers, o-NB side chain functionalization for thin film patterning, o-NB for self-assembled monolayers, photo cleavable block copolymers, and photo cleavable bioconjugates.
Solubility
Soluble in water 5000 mg/L 20 deg Celsius.
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
The o-nitrobenzyl group (o-NB) is utilized frequently in polymer and materials science. It covers the use of based cross-linkers for photodegradable hydrogels, o-NB side chain functionalization in (block) copolymers, o-NB side chain functionalization for thin film patterning, o-NB for self-assembled monolayers, photo cleavable block copolymers, and photo cleavable bioconjugates.
Solubility
Soluble in water 5000 mg/L 20 deg Celsius.
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications
Melting Point | 70°C to 74°C |
Boiling Point | 270°C |
Quantity | 25 g |
Beilstein | 2046649 |
Sensitivity | Light sensitive |
Solubility Information | Soluble in water 5000mg/L 20 deg Celsius. |
Formula Weight | 153.14 |
Percent Purity | 97% |
Chemical Name or Material | 2-Nitrobenzyl alcohol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'