Få mere at vide
2-Nitrobenzoic-d4 Acid, CDN
Artikelnummer. 30317338
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317338 0.1 g
30311501 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317338 Leverandør CDN Leverandørnr. CDND73160.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Nitrobenzoic Acid-D4
CAS 855997-63-6
Molekylær formel C7 D4 H N O4
Mængde 0.1 g
Synonym Benzoic-2,3,4,5-d4 acid, 6-nitro- (9 CI), 6-Nitrobenzoic-2,3,4,5-d4 acid, 2-Nitrobenzoic-d4 Acid, o-Nitrobenzoic Acid-d4, 2-Nitrobenzoic Acid-D4
InChI formel InChI=1 S/C7H5NO4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h1-4 H,(H,9,10)/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c([2 H])c(c(C(=O)O)c1[2 H])[N+](=O)[O-]
IUPAC navn 2,3,4,5-tetradeuterio-6-nitrobenzoic acid
Molekylvægt (g/mol) 171.1436
Formel vægt 171.047 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity