Få mere at vide
2-Nitrobenzene-1,4-diol, TRC
Artikelnummer. 30427236
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30427236 100 mg
30413855 50 mg
30427326 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30427236 Leverandør Toronto Research Chemicals Leverandørnr. TRCN900973100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 16090-33-8
Molekylær formel C6H5NO4
Molekylvægt (g/mol) 155.11
IUPAC navn 2-nitrobenzene-1,4-diol
SMIL Oc1ccc(O)c(c1)[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 2-Nitrobenzene-1,4-diol
Mængde 100 mg
InChI formel InChI=1S/C6H5NO4/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,8-9H
Formel vægt 155.0219